Ti disc Embedding Ring, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
55110-TIR
102092-804PK 163.56 USD
102092-804
Ti disc Embedding Ring, Electron Microscopy Sciences
Embedding Cassettes Embedding Rings
Ti disc Embedding Ring for TEM sample preparation. Used for the most difficult step of cross-sectional thinning is the mechanical preparation of the sample, cutting a cross section and then embedding it into an appropriately designed ring.
Using this ring, embedding cross sections is greatly simplified, since the samples do not need to be glued together prior to embedding. In order to fix the samples in the ring it is deformed mechanically by an appropriate tool at the two points marked by the arrows. This deformation will both hold the sample in the ring and press both sample pieces together before gluing. Ti disc is also offered for embedding bulk samples of brittle material for plan-view TEM investigations.
Delivery information: The rings supplied in 5 sizes 0.6, 0.8, 1.0, 1.2, and 1.3 mm.