Kwik-Gard™ Sylgard 184 Silicone, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
24236-01 24236-02 24236-03 24236-04
102092-304KT 535.56 USD
102092-304 102092-306 102092-308 102092-310
Kwik-Gard™ Sylgard 184 Silicone, Electron Microscopy Sciences
Adhesives
The Kwik-Gard™ is specially packaged Sylgard184 silicone that offers a quicker and easier way to apply the silicone, eliminating the messy procedure of preparing the mixture before application.
  • Quicker and easier way to apply Sylgard184 silicone
  • Eliminates messy preparation
  • Does not need degassing for most applications
The special cartridge controls the precise mixing ratio to ensure proper curing. The disposable tip mixes resin and hardener as they are dispensed. Since no air is introduced during mixing, the resin does not need degassing for most applications. The mixed silicone is applied directly to the site, reducing preparation time and material waste. Each Kwik-Gard™ cartridge contains 37 mL of resin and hardener. The dispensing tip has a dead volume of 0.75 mL. Curing time for this silicone is 24 hours at 25°C, 15 minutes at 150°C.