Blue Silicone Rubber, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
24230-B
102092-300EA 378.47 USD
102092-300
Blue Silicone Rubber, Electron Microscopy Sciences
Specimen Mounting Supplies
Blue silicone rubber is extremely high tear, high tensile strength, two components and tin catalyzed RTV silicone rubber
  • Low shrinkage and low viscosity
  • Excellent chemical resistance
  • Excellent shelf aging
  • Color coded catalyst
  • Long pot life insensitive to inhibition
  • Variable cure rate
  • Good dielectric properties
It is designed for casting polyurethane foam, polyester and epoxy parts.
Delivery information: This kit includes 1 gal (9 lbs.) silicone base and 1 pt (1 lbs.) blue catalyst.