Formalin 2%, (non-buffered)

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol

15735-01
102091-854EA 48.73 USD
102091-854
Formalin 2%, (non-buffered)
Formalin
Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR