Dumont Tweezers Style N0C, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0304-N0C-PO
102091-704EA 56.7 USD
102091-704
Dumont Tweezers Style N0C, Electron Microscopy Sciences
Forceps
Type 0 short, thin tips, 108 mm.
Antimagnetic is composed of C, Cr, Ni and Mo. It offers a good resistance to corrosion thanks to its high concentration of molybdenum and is 80% antimagnetic but is not as hard as Inox. Antimagnetic is resistant up to temperatures of around 400°C (DIN 50 914) and is suitable for autoclave sterilization at 270°C.