EMS Ultramicrotome Aid, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70612
102091-154EA 513.43 USD
102091-154
EMS Ultramicrotome Aid, Electron Microscopy Sciences
Microtomes and Microtome Assemblies Microtome Blades and Handles
Ultramicrotome Aids are made from hard plastic (Delrin) with a beautifully machined finish to handle many of your instruments that need to be at your fingertips during sectioning.
This EMS Ultra-microtome Aid is a safe place to rest your diamond knife, diamond  trimming tool, tweezers, hair curl or eye lash, diamond knife cleaning rods, perfect-loop for picking up sections.
Caution: Note: This unit is not a safe device to store your sectioning knives, such as diamond or glass knives, after you’re done the sectioning.