Bacterial Cell Spreader, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62452
102091-068PK 153.57 USD
102091-068
Bacterial Cell Spreader, Electron Microscopy Sciences
Spreaders Cell Spreaders
Made from high impact polystyrene. No flame sterilization needed. Designed for easy spreading of cells onto the surface of an agar plate. Supplied sterile in individually packs. Total length of handle: 176 mm, width of spreader: 36 mm. Color green.