Acrytol Mounting Medium, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
13518
102090-972EA 78.94 USD
102090-972
Acrytol Mounting Medium, Electron Microscopy Science
Stains Microscopy and staining reagents
Acrytol is a rapid-drying mounting medium which enables slides to be screened immediately. It has a low viscosity, allowing the medium to flow easily. It also prevents air bubbles from becoming trapped.
Acrytol contains an antioxidant to inhibit stain fading and prevent the formulation of annual rings. A thin coating of Acrytol is all that is needed to adhere a coverglass to a microscope slide.