Sodium chloride 1.6% in 80% alcohol

Supplier: Electron Microscopy Sciences

10381
102090-960EA 69.49 USD
102090-960
Sodium chloride 1.6% in 80% alcohol
Sodium chloride

Alkaline Alcohol formulations are 10% NH4OH in 85.5% alcohol, 1.6% NaCl in 80% alcohol, and 0.2% KOH in 80% alcohol.

Formula: NaCl
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR