Anti-CD3 Epsilon Polyclonal Antibody

Supplier: Boster Biological Technology
PB9093
10209-316EA 912.63 USD
10209-316
Anti-CD3 Epsilon Polyclonal Antibody
Antibodies
Polyclonal antibody for CD3/CD3E detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CD3/CD3E information: Molecular Weight: 23147 MW; Subcellular Localization: Membrane; Single-pass type I membrane protein.

Type: Primary
Antigen: CD3 Epsilon
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR