Anti-PCK-26 Mouse Monoclonal Antibody [clone: PC-26]

Supplier: Boster Biological Technology
MA1082
10209-136EA 912.68 USD
10209-136
Anti-PCK-26 Mouse Monoclonal Antibody [clone: PC-26]
Antibodies
Mouse IgG monoclonal antibody for PCK-26 detection. Tested with WB, IHC-P, IHC-F in Human;rat. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: PCK-26
Clonality: Monoclonal
Clone: PC-26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR