Anti-GRIA3 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA2249
10209-134EA 912.68 USD
10209-134
Anti-GRIA3 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Glutamate receptor 3(GRIA3) detection. Tested with WB in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: Glutamate receptor 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR