Anti-IgG Antibody (HRP (Horseradish Peroxidase))

Supplier: Boster Biological Technology
BA1050-1
10208-922EA 527.28 USD
10208-922
Anti-IgG Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR