Anti-Thrombospondin 2 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1417
10207-956EA 911.73 USD
10207-956
Anti-Thrombospondin 2 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Thrombospondin-2(THBS2) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: Thrombospondin 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR