Anti-KDR Mouse Monoclonal Antibody [clone: FLK-6]

Supplier: Boster Biological Technology
MA1040
10207-940EA 911.73 USD
10207-940
Anti-KDR Mouse Monoclonal Antibody [clone: FLK-6]
Antibodies
Mouse IgG monoclonal antibody for VEGFR2, kinase insert domain receptor (a type III receptor tyrosine kinase) (KDR) detection. Tested with WB, IHC-F in Human. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: KDR
Clonality: Monoclonal
Clone: FLK-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR