Anti-CDC25C Mouse Monoclonal Antibody [clone: IMD-25]

Supplier: Boster Biological Technology
MA1018
10207-936EA 911.75 USD
10207-936
Anti-CDC25C Mouse Monoclonal Antibody [clone: IMD-25]
Antibodies
Mouse IgG monoclonal antibody for CDC25C, cell division cycle 25 homolog C (S. pombe) (CDC25C) detection. Tested with WB in Human. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: CDC25C
Clonality: Monoclonal
Clone: IMD-25
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR