Mouse PCSK9 PicoKine ELISA Kit, Boster
Supplier: Boster Biological Technology
EK1148
10207-860EA 1229.56 USD
10207-860
Mouse PCSK9 PicoKine ELISA Kit, Boster
Assays ELISAs
Sandwich ELISA kit of Quantitative Detection for Mouse PCSK9
Ordering information: Kits include 96-well plate precoated with anti- mouse PCSK9 antibody 1; ST0000-4 Lyophilized mouse PCSK9 standard 4ng/tubex2; AR1107 Biotinylated anti- mouse PCSK9 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.