Anti-IgG Antibody (Biotin)

Supplier: Boster Biological Technology
BA1003-0.5
10207-520EA 275.32 USD
10207-520
Anti-IgG Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR