Anti-IL3 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1748
10207-506EA 912.68 USD
10207-506
Anti-IL3 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Interleukin-3(IL3) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: IL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR