Anti-MAP2K6 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1921
10207-320EA 912.68 USD
10207-320
Anti-MAP2K6 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Dual specificity mitogen-activated protein kinase kinase 6(MAP2K6) detection. Tested with WB in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: MAP2K6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR