Anti-SOCS1 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1074
10206-770EA 911.73 USD
10206-770
Anti-SOCS1 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Suppressor of cytokine signaling 1(SOCS1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: SOCS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR