Anti-TGF alpha Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1360
10206-474EA 911.75 USD
10206-474
Anti-TGF alpha Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Protransforming growth factor alpha(TGFA) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: TGF alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR