Anti-CDH5 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA2079
10206-390EA 923.79 USD
10206-390
Anti-CDH5 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Cadherin-5(CDH5) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: CDH5
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR