Anti-Caspase-1(P20) Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1440
10206-318EA 912.68 USD
10206-318
Anti-Caspase-1(P20) Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Caspase-1(CASP1) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: Caspase-1(P20)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR