Anti-ALP Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1004
10206-078EA 912.44 USD
10206-078
Anti-ALP Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Alkaline phosphatase, tissue-nonspecific isozyme(ALPL) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: ALP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR