Anti-GH1 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
RP1024
10206-076EA 912.63 USD
10206-076
Anti-GH1 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Somatotropin(GH1) detection. Tested with WB in Mouse.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: Growth Hormone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR