Anti-Insulin Mouse Monoclonal Antibody [clone: ISL-8J]

Supplier: Boster Biological Technology
MA1052
10206-062EA 912.68 USD
10206-062
Anti-Insulin Mouse Monoclonal Antibody [clone: ISL-8J]
Antibodies
Mouse IgG monoclonal antibody for Insulin, insulin (INS) detection. Tested with IHC-P in Human;mouse;rat. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: Insulin
Clonality: monoclonal
Clone: ISL-8J
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR