Anti-API5 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1009
10206-038EA 912.68 USD
10206-038
Anti-API5 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Apoptosis inhibitor 5(API5) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: API5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR