Anti-Tenascin Mouse Monoclonal Antibody [clone: T20]

Supplier: Boster Biological Technology
MA1094
10206-030EA 912.68 USD
10206-030
Anti-Tenascin Mouse Monoclonal Antibody [clone: T20]
Antibodies
Mouse IgG monoclonal antibody for Tenascin, tenascin N (TNN) detection. Tested with WB, IHC-P, IHC-F, ICC in Human. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: Tenascin
Clonality: monoclonal
Clone: T20
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR