Mouse Galectin-7 PicoKine ELISA Kit, Boster
Supplier: Boster Biological Technology
EK1279
10205-434EA 1228.1 USD
10205-434
Mouse Galectin-7 PicoKine ELISA Kit, Boster
Assays ELISAs
Sandwich ELISA kit of Quantitative Detection for Mouse Galectin-7
Ordering information: Kits include 96-well plate precoated with anti- mouse Galectin-7antibody 1; ST0000-10 Lyophilized recombinant mouse Galectin-7 standard 10ng/tubex2; AR1107 Biotinylated anti- mouse Galectin-7 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex (ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.