Vented Paclitaxel Set with Polyethylene Lined Tubing, INTERLINK Injection Site
Supplier: Baxter
2C7557
10205-172CS 3805.57 USD
10205-172
Vented Paclitaxel Set with Polyethylene Lined Tubing, INTERLINK Injection Site
Valves Injection Valves
Vented paclitaxel set with polyethylene lined tubing, INTERLINK injection site with 0.22 µm downstream high pressure filter, non-DEHP 12" pump segment.
  • Male Luer lock adapter
  • Contains DEHP
  • Approximately 10 drops/ml
  • Fluid path is sterile, nonpyrogenic