Sodium chloride 0,9% USP for injection

Supplier: BAXTER IV SOLUTIONS

Synonyms: Saline, NaCl, Sea salt, Halite

2B0042
10205-168CS 2067.6 USD
10205-168
Sodium chloride 0,9% USP for injection
Sodium chloride

Sterile solutions for injection.


  • Intravenous use
  • Subcutaneous use
  • mEq/L: Na 154, Cl 154
  • mOsmol/L (calc): 308
  • NDC Number: 0338-0553-11
  • pH: 5.5 (5.0–7.5)


Packaging: Multi Pack contains 16 bags per overwrap pouch. Quad Pack contains four bags per overwrap pouch. Single Pack contains one bag per overwrap pouch.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR