Speedglas™ Spacer Bar, 3M™
Supplier: 3M
7100031975
10204-934CS 12.61 USD
10204-934
Speedglas™ Spacer Bar, 3M™
Welding Supplies
Speedglas™ Spacer Bar helps prevent the inside protection plate from rubbing on the quick-release rail as it slides up and down during use.
The Spacer Bar is designed for use with Speedglas™ Auto-Darkening Filters 9100X/9100XX.