SP Bel-Art Stainless Steel Clamping Thermometer Holder, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B61400-7300
10204-914CS 273.28 USD
10204-914
SP Bel-Art Stainless Steel Clamping Thermometer Holder, Bel-Art Products, a part of SP
Tool Sets
With five hole sizes ranging from 3.5 mm to 14.5 mm, this holder accommodates various thermometers including liquid in glass, electronic with probe, and bimetal with probe.
  • Stainless steel construction
  • Durable polyurethane foam keeps thermometers securely in place
  • Rubber bumpers prevent scratching
The stainless steel, spring-loaded clip attaches securely to laboratory containers, shelves up to 3/8" thick, and more. Rubber bumpers on the clip prevent scratching. Durable polyurethane foam provides a secure grip to keep thermometers in place.