Tissue-Tek® AutoSection® Automated Microtome, Sakura Finetek
Supplier: Sakura Finetek
5010
10204-886EA 41680.25 USD
10204-886
Tissue-Tek® AutoSection® Automated Microtome, Sakura Finetek
Microtomes and Microtome Assemblies Microtomes
AutoSection automatically aligns block to the blade edge, saving tissue and technician time.
  • Auto-aligns block to blade edge, even if block was previously sectioned on a different microtome
  • One touch trimming and sectioning elimate repetitive hand wheel motions and reduces the risk of musculoskeletal disorders
  • 3D chuck/specimen holder
  • LCD touch screen control panel
  • Wireless remote control
  • One-touch operation to engage cutting modes
  • 16 user-designed sectioning programs
Tissue-Tek® AutoSection® is the most precise microtome on the market as determine by lasar measurements
Multiple built-in safeguards make AutoSection® the leader in microtome user safety. One-touch trimming and sectioning eliminate repetitive handwheel motions and reduces the risk of musculoskeletal disorders.
Programmable sectioning enables laboratories to implement standardized protocols for specific tissue or block types. AutoSection® also standardizes section thickness to help optimise advanced staining procedures such as IHC and ISH. Patented AutoAlign and AutoTrim features assure preservation of tissue as well as saving technician's time.