MPure™ Total RNA Extraction Kit, MP Biomedicals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

117022160
10204-748EA 645.22 USD
10204-748
MPure™ Total RNA Extraction Kit, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

MPure Total RNA Extraction Kit is used with the MPure-12 instrument for extraction of total RNA from whole blood, blood cells, animal tissue, plant tissue, yeast or cultured cells.


  • Sample Type: Whole Blood, Yeast, Cultured Cells, Animal Tissue, Plant Tissue, Blood Cells
  • Sample Volume: 100-400 µL


Application Notes
Nucleic acids extracted can be used in a number of downstream applications including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP, Southern Blot Analysis.


Usage Statement
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.


Key Applications
Total RNA extracted from MPure-12 Total RNA Extraction kit can be used in a number of downstream application including: RT-PCR; qPCR; Sequencing(NGS); Microarray; DDRT PCR; TaqMan® Analysis; Northern Blot AnalysisNorthern blotting.


Packaging: Kit contains Kit contains Proteinase K Solution = 40 µl, Lysis Buffer 2 = 720 µl, Binding Buffer 1 = 1300 µl, Magnetic Bead Solution = 800 µl, Washing Buffer 1 = 1000 µl, Washing Buffer 2 = 1000 µl, Washing Buffer 3 = 1000 µl, RNase Free Water = 1000 µl, Rnase Free Water = 1000 µl, DNase Buffer = 1000 µl

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR