MPure Plant DNA Extraction Kit, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
117022150
10204-746EA 645.22 USD
10204-746
MPure Plant DNA Extraction Kit, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
MPure Plant DNA Extraction Kit is used with the MPure-12 instrument for extraction of genomic DNA from plant (leaf, seeds and spores) and fungal tissues. Up to 100 mg of tissue can be used for purification.
  • Extraction of genomic DNA from plant and fungal tissues
  • Fresh or lyophilized plant tissue
  • Fully formed mycelium or cells from yeast culture
  • Completely automated purification protocol
  • Use with MPure-12 System
  • Run in as little as 40 minutes
  • DNA can be used immediately in downstream applications
MPure™ Plant DNA Extraction Kit contains everything needed for sample preparation. However, for tough tissues such as roots and seeds, or tough fungal tissue, sample preparation using FasrPrep™ Instruments and Lysing Matrix Tubes can greatly increase yields as well as shorten processing time.
Nucleic acids extracted from MPure-12 Plant DNA Extraction kit can be used in a number of downstream application including: PCR; qPCR; Sequencing(NGS); Microarray; RFLP; Southern Blot Analysis
Capacity: 400 µL
Sample Type: Fungal Tissues, Plant (leaf, seeds and spores)
Packaging: Kit contains Lysis Buffer 2 = 720 µl, Binding Buffer 1 = 720 µl, Magnetic Bead Solution = 800 µl, Washing Buffer 1 = 1000 µl, Washing Buffer 2 = 1000 µl, Washing Buffer 3 = 1000 µl, Elution Buffer 1 = 1000 µl, Elution Buffer 2 = 1000 µl
Caution: Kit should be stored at room temperature (15-25°C). Do not freeze the reagent cartridges. The Kits are stable for 24 months under these conditions. Store the purified DNA at 4°C (short- term) or aliquot and store at -70°C (long-term) before performing the downstream analysis.