FCS2®/FCS3® 30-Piece Gasket Set, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
06031920719
10200-066PK 415.07 USD
10200-066
FCS2®/FCS3® 30-Piece Gasket Set, Bioptechs Inc.®
Seals Gaskets
A 30-piece assortment of round gaskets, featuring 14×22 mm and 30 mm round internal cutouts plus blanks of each thickness.
  • Unlimited options for internal geometry
  • Creates user definable flow characteristics
  • Enables near laminar flow
Designed for use with FCS2® or FCS3® chambers.
Certifications: CE certified.