CFCS2® Cooled Closed Chamber System, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
030603192
10200-026EA 6917.71 USD
10200-026
CFCS2® Cooled Closed Chamber System, Bioptechs Inc.®
Flow Cells
This modified FCS2® Closed Chamber System includes fluid ports in the base to accommodate the circulation of an external refrigerant.
  • Suitable for no flow through high rate flow procedures where a rapid exchange of media is required with low cell surface shear
  • Closed system so that bicarbonate CO2 or organic buffers can be employed
  • Compatible with 1/16" tubing for perfusion (C-Flex, Tygon, etc.)
  • Easily assembled with no tools required
  • Unlimited options for internal geometry
  • Creates user definable flow characteristics
  • Enables near laminar flow
Certifications: CE certified.
Ordering information: System includes Chamber, five Non-Heating Microaqueduct Slides, fifty Coverslips, and 30-piece Gasket Set. This chamber has heating and cooling capabilities, but does not include temperature controller.