FCS2® Cooling Adapter, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
06031920301
10200-020EA 1106.82 USD
10200-020
FCS2® Cooling Adapter, Bioptechs Inc.®
Fittings and Connectors
The FCS2® Cooling Adapter is an O-Ring sealed window that circulates an isolated cooled fluid over the upper glass element (Microaqueduct slide) to cool the specimen to desired temperature
This Cooling Adapter is designed for use with FCS2® Closed Chamber Systems.
Certifications: CE certified.