FCS2® Non-Heated Flow Cell, Bioptechs®
Supplier: BIOPTECHS INC.
030603192NH
10200-010KT 4980.78 USD
10200-010
FCS2® Non-Heated Flow Cell, Bioptechs®
Flow Cells
A parallel plate, high N.A
  • Near laminar flow perfusion
  • User definable flow characteristics
  • Compatible with all modes of microscopy including high NA/high magnification uses
  • Over 20 years of being the foremost chamber in micro-environmental control
  • Ideal for both flow and static applications
Compatible, microscopic flow cell with a user definable specimen area.
Certifications: CE certified.
Delivery information: Includes Non-Heated FCS2® Chamber, five non-heating micro aqueduct slides, one hundred #1.5 thickness, 40 mm German Desag263 glass Coverslips, and a 30-piece Gasket Set.