Perfusion/Temperature USB Controller, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
1316160313
10200-002EA 6821.78 USD
10200-002
Perfusion/Temperature USB Controller, Bioptechs Inc.®
Software Laboratory Software
A USB interfaced computer program and associated cabling designed to interface micro-perfusion pumps and Bioptechs temperature controllers
  • Dual or single micro-perfusion pumps
  • Flow profile to reduce dead volume delays
  • Temperature and perfusion recording
  • Data logging of perfusion and temperature
  • Multi flow-rate calibration
  • Temperature profiling and cycling
  • Saving and reloading settings
  • Graphic display of events
The program is compatible with Windows XP, Windows 7, and Windows 8 systems. The program provides an intuitive means of control for perfusion or temperature in time lapse micro-imaging.
Certifications: CE certified.
Ordering information: Software only.