25-OH Vitamin D, Total ELISA, Gold Standard Diagnostics
Supplier: Gold Standard Diagnostics
KAP1971
10199-926KT 1784.91 USD
10199-926
25-OH Vitamin D, Total ELISA, Gold Standard Diagnostics
Assays ELISAs
Vitamin D testing offers important information for general physicians and specialists alike
This enzyme linked immunosorbent assay (ELISA) is intended for the quantitative determination of 25-hydroxyvitamin D2 and D3 (25-OH D2 and 25-OH D3). Intended for in vitro diagnostic use.
Key features include strong correlation with established LC/MS, ELISA, and CLIA methods, high precision, excellent sensitivity (2.8 ng/mL) and fast turnaround (<4 hours).
Long-established as an important factor for bone metabolism, new research indicates Vitamin D also plays an important role in immune function, cardiovascular disease, autoimmune conditions, diabetes, depression and cancer prevention. More than 75% of the U.S. population has a deficiency of 25- Hyrdoxyvitamin D.
Certifications: 510(k) cleared.
Ordering information: 96-well microtiter plate, 2 controls and 6 calibrators are provided.