Glass Coverslips, 30mm, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
3013130319
10199-874PK 62.42 USD
10199-874
Glass Coverslips, 30mm, Bioptechs Inc.®
Cover Glasses
Low alkaline, German Desag 263 glass coverslips are optimized for cell culture growth
Coverslips feature a 30mm diameter and #1.5 thickness.
Ordering information: Pack includes 100 coverslips.