Objective Wrench and Plier Set, Bioptechs
Supplier: BIOPTECHS INC.
021523
10199-798KT 587.88 USD
10199-798
Objective Wrench and Plier Set, Bioptechs
Wrenches
This tool set used to isolate the mechanical stress applied to an objective when removing the decorative collar.
  • Safely allows removal of decorative collar
  • For use with the Objective Heater® System
Designed to enable the attachment of an objective heater.
Certifications: CE certified.