Oasis® Heating and Cooling System, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
15160
10199-782EA 5936.61 USD
10199-782
Oasis® Heating and Cooling System, Bioptechs Inc.®
Circulating Thermostats Closed Circulator Systems Temperature Control Systems
The Oasis® Heating and Cooling System is a Peltier-driven heating and cooling system that circulates temperature-regulated fluid through accessory devices used on a microscope.
  • Lightweight and portable
  • User-friendly interface
  • Universal power input
  • Low power consumption
  • Solid-State technology (no compressor)
  • Environmentally friendly
  • Leak-free disconnects
  • Quiet operation
  • Serial communication
  • Non-volatile memory set temperature
  • Wide temperature range of operation
Certifications: CE certified.
Ordering information: The Oasis® Heating and Cooling System requires tubing and cooling adapter for specific cooling.