Micro-Perfusion Pump Mounting Rod Clamp, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
16181303
10199-778EA 70.41 USD
10199-778
Micro-Perfusion Pump Mounting Rod Clamp, Bioptechs Inc.®
Clamps General Purpose Clamps
A polymer screw-on clamping bracket to support the Bioptechs Micro-Perfusion Pump onto conventional 1.27cm (0.5") breadboard support post.
Certifications: CE certified.