Pre-washed Pre-Weighed Glass Microfiber Filter, Paper Grade 169, Ahlstrom
Supplier: Ahlstrom
169W-0470
10199-602PK 179.84 USD
10199-602
Pre-washed Pre-Weighed Glass Microfiber Filter, Paper Grade 169, Ahlstrom
Filters Glass Fiber Filters
Grade 169 is a microfiber glass grade formulated for product stability when heated over 500 °C as required for testing for volatile solids in STM 2540.
In this item each filter is washed and weighed and comes in an aluminum pan with the weight printed on the side for ease of use.