Dissipative Label Dispenser, Desco
Supplier: Desco
06767
10199-584EA 6.66 USD
10199-584
Dissipative Label Dispenser, Desco
Label and Tape Dispensers
Dissipative Label Dispenser helps peel away backing from label. Designed for use with labels up to 2" wide.
  • Volume Resistance: Rv 1×10E6 to < 1×10E9 ohms per ANSI/ESD STM11.12
  • Made in the United States of America
Designed for used in an ESD protected area. Place on a grounded ESD work surface to ground. Label rolls must have a core to work with the label dispenser.
Ordering information: Item ships unassembled.