Debakey Pediatric Forceps, Sklar
Supplier: Sklar
52-1904
10199-370EA 504.74 USD
10199-370
Debakey Pediatric Forceps, Sklar
Forceps
Designed for use on pediatric patients. Forceps are commonly used for grasping fine tissue and blood vessels or soft tissue dissection. The narrow tips and atraumatic teeth cause little to no damage to the tissues.
  • Nontraumatic fine teeth: 1×2 rows
  • Finger ring handles
  • Reusable
  • Latex-free